missing translation for 'onlineSavingsMsg'
Learn More

Sodium chlorite, tech. nominally 80%

Product Code. 11327857
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
Unit Size:
10kg
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327857 10 kg 10kg
11317857 500 g 500g
11337857 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11327857 Supplier Thermo Scientific Alfa Aesar Supplier No. 014265.A9
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium chlorite, ClNaO3, CAS Number-7758-19-2, 7775-09-9, 1310-73-2, 497-19-8, 7647-14-5, 7757-82-6, chlorate de sodium, asex, chloric acid, sodium salt, atlacide, tumbleleaf, rasikal, kusatol, chlorsaure, desolet, sodium chlorate, 10kg, White flake/powder, Nominally 80%, CHEBI:65242 | CAS: 7758-19-2 | ClNaO3 | 106.44 g/mol

Sodium chlorite is used in the in situ generation of chlorine dioxide for stripping of textiles, bleaching, pulp and paper industries. It acts as a disinfectant in water treatment plant and as a preservative in eye drops. It is also used in contact lens cleaning solution and for sanitizing air ducts. It is associated with zinc chloride and used as a component in therapeutic rinses, toothpastes, mouth sprays and gels. It is utilized for the synthesis of 4-oxo-2-alkenoic acids from alkyl furans. Further, it is involved as a reagent in Pinnick oxidation reaction to prepare carboxylic acid from aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-19-2 , 7775-09-9 , 1310-73-2 , 497-19-8 , 7647-14-5 , 7757-82-6
MDL Number MFCD00003478
IUPAC Name sodium;chlorite

Specifications

Melting Point 180°C to 200°C (decomposition)
Color White
Physical Form Flake or Powder
Quantity 10 kg
Assay Percent Range Nominally 80%
Molecular Formula ClNaO3
Synonym sodium chlorite, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
Solubility Information Soluble in water. Slightly soluble in methanol and ethanol.
InChI Key YZHUMGUJCQRKBT-UHFFFAOYSA-M
SMILES [Na+].[O-][Cl](=O)=O
Molecular Weight (g/mol) 106.44
PubChem CID 516902
ChEBI CHEBI:65242
Formula Weight 90.44
Grade Technical
Concentration or Composition (by Analyte or Components) Sodium chlorite: 80.0%; Sodium chlorate: 3.0%; Sodium hydroxide: 3.0%; Sodium carbonate: 2.0%; Sodium chloride: 10.0%; Sodium sulphate: 0.3%; Water: 1.7%
Sensitivity Hygroscopic
Appearance White flake/powder
Chemical Name or Material Sodium chlorite
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.