missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chlorite, Honeywell Fluka™

Product Code. 15692640 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
Unit Size:
100g
1kg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15692640 1 kg 1kg
15602650 5 g 5g
15682640 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15692640 Supplier Honeywell-Fluka Supplier No. 2441551KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Technical Grade, 80%

  • Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
  • Variation in color and physical form is possible
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-19-2
Molecular Formula ClNaO2
Molecular Weight (g/mol) 90.438
MDL Number MFCD00003478
InChI Key UKLNMMHNWFDKNT-UHFFFAOYSA-M
Synonym sodium chlorite, chlorous acid, sodium salt, textone, chlorite sodium, textile, alcide ld, neo silox d, caswell no. 755, sodiumchlorite, unii-g538ebv4vf
PubChem CID 23668197
ChEBI CHEBI:78667
IUPAC Name sodium;chlorite
SMILES [O-]Cl=O.[Na+]

Specifications

Quantity 1 kg
Linear Formula NaClO2
Formula Weight 90.44g/mol
Grade Technical
Concentration or Composition (by Analyte or Components) 80%
Chemical Name or Material Sodium Chlorite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.