missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chlorite, Anhydrous, Flakes, BAKER™, J.T. Baker™

Product Code. 15528834 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15528834 500 g Wide Mouth Amber Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15528834 Supplier Avantor J.T.Baker Supplier No. V04407

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A grade of chemicals for laboratory use

Specifications

CAS 7758-19-2
Physical Form Flakes
Quantity 500 g
Molecular Formula ClNaO2
Synonym sodium chlorite, chlorous acid, sodium salt, textone, chlorite sodium, textile, alcide ld, neo silox d, caswell no. 755, sodiumchlorite, unii-g538ebv4vf
InChI Key UKLNMMHNWFDKNT-UHFFFAOYSA-M
SMILES [O-]Cl=O.[Na+]
IUPAC Name sodium;chlorite
Molecular Weight (g/mol) 90.438
PubChem CID 23668197
ChEBI CHEBI:78667
Formula Weight 90.44
Packaging Wide Mouth Amber Glass Bottle
Chemical Name or Material Sodium Chlorite, Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.