missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride USP/FCC Granular, Macron Fine Chemicals™

Product Code. 15588414
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly
Unit Size:
2.5kg
This item is not returnable. View return policy

Product Code. 15588414

Brand: Avantor Macron 753206

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Physical Form Granules
Quantity 2.5 kg
IUPAC Name sodium;chloride
Formula Weight 58.44
Grade FCC/USP
Packaging Poly
Chemical Name or Material Sodium Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.