Learn More
Sodium chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Sodium Chloride is widely used in biochemistry and molecular biology applications. It is used in a wide variety of biochemical applications including creating density gradients, regulating biological functions and as a diluent to increase ionic strength in buffers or culture media, a component of PBS and SSC buffers. It has also been used for precipitation of DNA from SDS-containing samples and in the removal of small nucleic acid fragments from plasmid DNA preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Boiling Point | 1413°C |
Melting Point | 801°C |
Color | White |
pH | 6 to 9 |
Physical Form | Beads |
Quantity | 5 g |
Merck Index | 14,8599 |
Solubility Information | Soluble in water (358mg/ml at 20°C),alcohol (very slightly),glycerol (100mg/ml),and ammonia. Insoluble in hydrochloric acid. |
IUPAC Name | sodium;chloride |
Formula Weight | 58.44 |
Show More |
RUO – Research Use Only