missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, ultra dry, 99.99% (metals basis)

Product Code. 11338567
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338567 100 g 100g
11425050 5 g 5g
11348567 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11338567 Supplier Thermo Scientific Alfa Aesar Supplier No. 035716.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7647-14-5 | ClNa | 58.44 g/mol

Sodium chloride is widely used as a food additive, and a preservative. It is employed in the Chloralkali process for producing chlorine and sodium hydroxide, in the Solvay process for producing sodium carbonate and calcium chloride, in the Mannheim process and in the Hargreaves process for producing sodium sulfate and hydrochloric acid. It is used in the manufacture of sodium chlorate, water softening and de-icing of roadways. Further it is used in several industries like pharmaceutical, textile, dyeing, rubber food and agriculture. It is used in the processing of steel, copper, vanadium, and other metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1413°C
Melting Point 801°C
Color White
pH 6 to 9
Physical Form Beads
Quantity 100 g
Merck Index 14,8599
Solubility Information Freely soluble in water. Slightly soluble in alcohol and ammonia.
Formula Weight 58.44
Percent Purity 99.99%
Odor Odorless
Packaging Ampule
Refractive Index 1.5442
Sensitivity Hygroscopic
Density 2.165 g/mL
Assay (metals basis)
Chemical Name or Material Sodium chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.