missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride-Tris-EDTA (STE) Buffer, 1X (pH 8.0), Fisher BioReagents

Brand:  Fisher Bioreagents BP2478-1

Code : 32

Additional Details : Weight : 1.05000kg

Quantity 1L
Packaging Nalgene Poly Bottle

 View more versions of this product

Product Code. 10706844

  • £62.93 / 1 L


EU Stock 4
Add to basket

Description

Description

  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X STE buffer solution
  • Eliminates the hassle of dilution or waiting for powder to dissolve
  • 1X solution contains 10mM tris, 1mM EDTA, and 100mM NaCl
  • Filtered through a 5 micron filter and autoclaved
  • Tested for the absence of DNase, RNase, and protease
  • STE buffer is used for DNA extraction protocols
  • Intended for laboratory research use only
Specifications

Chemical Identifiers

Sodium Chloride-Tris-EDTA Buffer
1X, pH 8.0
7732-18-5
Saline Tris–EDTA, STE, TEN

Specifications

Sodium Chloride-Tris-EDTA Buffer
7732-18-5
DNase free
Nalgene Poly Bottle
Filtered through a 5-micron filter.
8
1L
1X, pH 8.0
Liquid
Saline Tris–EDTA, STE, TEN
Protease free
100°C
Clear
Videos
SDS Documentation