missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, TraceSELECT™, for trace analysis, ≥99.999% (metals basis), Honeywell Fluka™

Product Code. 16635853 Shop All Honeywell Fluka Products
Change view
Click to view available options
:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16635853 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16635853 Supplier Honeywell Fluka Supplier No. 38979500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Boiling Point 1,465°C (1,013 hPa)
Melting Point 800°C
CAS 7647-14-5
Color Colorless
CAS Max % ≤ 100%
pH 7
Quantity 500 g
Molecular Formula ClNa
Linear Formula NaCl
MDL Number MFCD00003477
Beilstein 3534976
Synonym Halite
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
Grade Trace Analysis
Density 2.160 g/cm3 (20°C)
Chemical Name or Material Sodium chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.