missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, ReagentPlus™, Honeywell™

Product Code. 15638140
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Quantity 10 kg
Linear Formula NaCl
UN Number NONH for all modes of transport
Beilstein 3534976
IUPAC Name sodium chloride
Formula Weight 58.44g/mol
Percent Purity ≥99%
Grade ReagentPlus™
Chemical Name or Material Sodium chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'