missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chloride, Puriss., meets analytical spec. of BP, Ph. Eur., USP, 99.0-100.5% (calc. to the dried substance), ≤0.00002% Al, Honeywell Fluka™
Shop All Honeywell Fluka Products

Click to view available options
Quantity:
1 kg
5 kg
10 kg
25 kg
32 kg
4 x 5 kg
6 x 1 kg
Unit Size:
10kg
1kg
25kg
32kg
5kg
Pack of 4
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 6 x 1 kg |
| Linear Formula | NaCl |
| UN Number | NONH for all modes of transport |
| IUPAC Name | sodium;chloride |
| Formula Weight | 58.44 |
| Percent Purity | 99.0 to 100.5% (calc. to the dried substance) |
| Grade | puriss. |
| Chemical Name or Material | Sodium Chloride, Meets analytical specification of Ph. Eur.,BP, USP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction