missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Sodium Chloride (NaCl) Solutions, 0.1–2.0 M

Product Code. 18074080
Click to view available options
Quantity:
5 L
20 L
200 L
Packaging:
Drum
Sample
Tote
Concentration:
0.1 M
0.2 M
0.5 M
1.0 M
2.0 M
Unit Size:
20 L
200 L
5 L
This item is not returnable. View return policy

Product Code. 18074080

Brand: Gibco™ A5803002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gibco Sodium Chloride (NaCl) solutions are sterile-filtered, packaged in bioprocess containers, and ready-to-use. Sourcing NaCl solutions can help streamline your buffer preparation allowing you to focus on additional critical steps in your biopharmaceutical processes.

We mix compendial-grade sodium chloride (NaCl) with the same bulk water used in our Gibco Water for Injection (WFI) for Cell Culture to produce solutions that meet the most stringent expectations of the pharmaceutical, biopharma, and diagnostic industries. NaCl solutions are manufactured to cGMP (21CFR820) and ISO 13485 standards, promoting consistency and convenience for every stage of the bioprocessing pipeline.

Product features:
• Multiple concentrations available (0.1 M, 0.2 M, 0.5 M, 1.0 M, 2.0 M)
• Available in a range of pack sizes (5 L, 20 L, 200 L)
• Established products with set specifications are ready to order
• Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
• Readily accessible documentation for accelerated qualification

User benefits:
• Eases the time requirements dedicated for in-house buffer preparation
• Mitigates space required for equipment storage
• Streamlines in-house chromatography processes related to formulation, hydration, testing, and validation
• Meets workflow needs for all stages of biologic development and manufacturing

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Chloride
Color Clear
Concentration 1.0 M
pH 4.5–8.0
Physical Form Liquid
Quantity 20 L
Formulation 1.0 M NaCl
Packaging Tote
Sterility Sterile-filtered
Certifications/Compliance GMP - 21 CFR 820, ISO 13485
Sterilization Method 0.2 μm Sterile-filtered
Endotoxin Level ≤ 1.0 EU/mL
Osmolality 1810–1937 mOsm/kg
Vis mere Vis mindre

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.