missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, MP Biomedicals™

Product Code. 11487872 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg
Unit Size:
10kg
1kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487872 1 kg 1kg
11477872 500 g 500g
11497872 5 kg 5kg
11407882 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11487872 Supplier MP Biomedicals Supplier No. 0215257591

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Commonly used in biochemical applications, including as a density gradient. Sodium Chloride, MP Biomedicals is found widely in nature and is considered to be an essential nutrient. Excess amounts can destroy electrolyte balance and cause death in most ani

A salt-ice mixture in the ratio of approximately 33g sodium chloride to 100g ice (at −1°C) will drop in temperature to as low as −21°C, depending on the rate of stirring and the size of the ice chunks.

Sodium chloride is used in a wide variety of biochemical applications, including:

  • Intravenous fluids (normal saline is 0.9% w/v in water)
  • Density gradients
  • A diluent to increase ionic strength in buffers or culture media
  • In salt-out procedures in the isolation of DNA
  • In high concentrations, for preservation of foods (since bacteria cannot grow in high salt conditions)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Physical Form Crystalline Powder
Quantity 1 kg
Solubility Information Soluble in water (357mg/mL at 25°C; 384mg/mL at 100°C), glycerol (100mg/mL); very slightly soluble in ethanol. Solubility in water is decreased by HCl; almost insoluble in concentrated HCl. Aqueous solutions has a neutral pH (6.7 - 7.3).
IUPAC Name sodium;chloride
Formula Weight 58.44
Grade ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.