missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
A salt-ice mixture in the ratio of approximately 33g sodium chloride to 100g ice (at −1°C) will drop in temperature to as low as −21°C, depending on the rate of stirring and the size of the ice chunks.
Sodium chloride is used in a wide variety of biochemical applications, including:
- Intravenous fluids (normal saline is 0.9% w/v in water)
- Density gradients
- A diluent to increase ionic strength in buffers or culture media
- In salt-out procedures in the isolation of DNA
- In high concentrations, for preservation of foods (since bacteria cannot grow in high salt conditions)
Specifications
Specifications
| Physical Form | Crystalline Powder |
| Quantity | 500 g |
| Solubility Information | Soluble in water (357mg/mL at 25°C; 384mg/mL at 100°C), glycerol (100mg/mL); very slightly soluble in ethanol. Solubility in water is decreased by HCl; almost insoluble in concentrated HCl. Aqueous solutions has a neutral pH (6.7 - 7.3). |
| IUPAC Name | sodium chloride |
| Formula Weight | 58.44 |
| Grade | ACS |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?