Learn More
Description
Sodium chloride is used in a wide variety of biochemical applications, including:
- Intravenous fluids (normal saline is 0.9% w/v in water10)
- Density gradients
- A diluent to increase ionic strength in buffers or culture media
- In salt-out procedures in the isolation of DNA
- In high concentrations for preservation of foods, since bacteria cannot grow in high salt conditions
A salt-ice mixture in the ratio of approximately 33g sodium chloride to 100g ice (at −1°C) will drop in temperature to as low as &minus’21°C, depending on the rate of stirring and the size of the ice chunks. Excess amounts of sodium chloride can destroy electrolyte balance and cause death in most animals, including humans.
Specifications
Specifications
| Physical Form | Crystalline Powder |
| Quantity | 5 kg |
| Solubility Information | Soluble in water (357mg/mL at 25°C; 384mg/mL at 100°C), glycerol (100mg/mL); very slightly soluble in ethanol. Almost insoluble in concentrated HCl. |
| IUPAC Name | sodium chloride |
| Formula Weight | 58.44g/mol |
| Percent Purity | ≥99.5% |
| Grade | Molecular Biology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.