missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, Free-flowing, Reagent Grade, anhydrous, ≥99%, Honeywell Fluka™

Product Code. 15637120 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
12 kg
25 kg
Packaging:
Fiberboard Box
Poly Bottle
Unit Size:
12kg
1kg
2.5kg
25kg
500g
5kg
This item is not returnable. View return policy

Product Code. 15637120

Brand: Honeywell-Fluka 793566500G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Free-flowing, Reagent Grade, anhydrous,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1,465°C
Melting Point 801°C (literature)
CAS Max % ≤100.0000%
Quantity 500 g
Linear Formula NaCl
Beilstein 3534976
IUPAC Name sodium chloride
Formula Weight 58.44
Percent Purity ≥99%
Grade Reagent
Packaging Poly Bottle
Density 2.160 g/cm3
Chemical Name or Material Sodium chloride, Free-flowing, Reagent Grade, anhydrous, ≥99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.