missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride crystal optic rectangle, 38.5mm x 19.5mm x 4mm, polished both sides, Thermo Scientific Chemicals

Product Code. 15454765
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7647-14-5
Quantity 1 Ea.
Molecular Formula ClNa
MDL Number MFCD00003477
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
PubChem CID 5234
ChEBI CHEBI:26710
Odor Odorless
Chemical Name or Material Sodium chloride crystal optic rectangle, 38.5mm x 19.5mm x 4mm, Polished both sides
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'