missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Boiling Point | 1465°C |
| Melting Point | 800°C |
| Color | Colorless |
| pH | 7.0 (at 20°C) |
| Physical Form | Crystalline |
| Quantity | 500 g |
| Linear Formula | NaCl |
| UN Number | NONH for all modes of transport |
| Beilstein | 3534976 |
| Solubility Information | 358g/L at 20°C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?