missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, 99.5%, for HPLC
GREENER_CHOICE

Product Code. 11964051 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Glass bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11964051 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11964051 Supplier Thermo Scientific Supplier No. 446212500
  • ONLINE EXCLUSIVE PRICE
    £105.00
    £94.65 / 250g
    Save £10.35
    9% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7647-14-5 | ClNa | 58.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 1461.0°C
Melting Point 801.0°C
CAS 7647-14-5
pH 5.0 to 9.0
Quantity 250 g
Assay Percent Range 99.4% min. (Argentometry)
Molecular Formula ClNa
Linear Formula NaCl
MDL Number MFCD00003477
Merck Index 15,8734
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
Solubility Information Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol (1g/10mL), ammonia, slightly soluble in alcohol, nearly insoluble in hydrochloric acid
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
PubChem CID 5234
ChEBI CHEBI:26710
Formula Weight 58.44
Percent Purity 99.5%
Grade HPLC
Packaging Glass bottle
Chemical Name or Material Sodium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.