missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chlorate, Technical, Fisher Chemical

Product Code. 10408230 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10408230 1 kg Plastic powder jar 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 10408230 Supplier Fisher Chemical Supplier No. S/3000/60

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

Melting Point 248°C
CAS 7775-09-9
Color White
Physical Form Solid
Quantity 1 kg
Molecular Formula ClNaO3
MDL Number 3479
Synonym sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
InChI Key YZHUMGUJCQRKBT-UHFFFAOYSA-M
SMILES [Na+].[O-][Cl](=O)=O
IUPAC Name sodium chlorate
Molecular Weight (g/mol) 106.44
PubChem CID 516902
ChEBI CHEBI:65242
Formula Weight 106.44g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium chlorate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H302-Harmful if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.