missing translation for 'onlineSavingsMsg'
Learn More

Sodium chlorate, ACS, 99.0% min

Product Code. 11458600
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458600 500 g 500g
11448600 100 g 100g
11468600 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11458600 Supplier Thermo Scientific Alfa Aesar Supplier No. 044408.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Maximum level of impurities: Insoluble matter 0.005%, BrO3 0.015%, Cl 0.005%, SO4 0.001%, Heavy metals (as Pb) 0.001%, Fe 5ppm, Ca 0.002%, Mg 0.002%, K 0.01% ∣ CAS: 7775-09-9 ∣ ClNaO3 ∣ 106.44 g/mol

Sodium chlorate is used to prepare chlorine dioxide in the pulp and paper industry. It is utilized for bleaching the wood pulp in order to make environmentally friendly and quality white paper products. It is also used to produce other perchlorate compounds in the industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-09-9
Molecular Formula ClNaO3
Molecular Weight (g/mol) 106.44
MDL Number MFCD00003479
InChI Key YZHUMGUJCQRKBT-UHFFFAOYSA-M
Synonym sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
PubChem CID 516902
ChEBI CHEBI:65242
SMILES [Na+].[O-][Cl](=O)=O

Specifications

Melting Point 248°C
Color Colorless
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99% min.
Solubility Information Soluble in water,glycerol,hydrazine,acetone and methanol. Slightly soluble in ethanol and ammonia.
IUPAC Name sodium chlorate
Formula Weight 106.45
Percent Purity ≥99%
Grade ACS
Odor Odorless
Density 2.5 g/cm3 at 20°C
Chemical Name or Material Sodium chlorate, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.