Learn More
Sodium chlorate, ACS, 99.0% min, Thermo Scientific™
Maximum level of impurities: Insoluble matter 0.005%, BrO3 0.015%, Cl 0.005%, SO4 0.001%, Heavy metals (as Pb) 0.001%, Fe 5ppm, Ca 0.002%, Mg 0.002%, K 0.01%
Brand: Thermo Scientific Alfa Aesar 044408.22
| Quantity | 100g |
|---|
View more versions of this product
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| ClNaO3 | |
| MFCD00003479 | |
| sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal | |
| CHEBI:65242 | |
| [O-]Cl(=O)=O.[Na+] |
Specifications
| 7775-09-9 | |
| 99% min. | |
| MFCD00003479 | |
| Soluble in water,glycerol,hydrazine,acetone and methanol. Slightly soluble in ethanol and ammonia. | |
| [O-]Cl(=O)=O.[Na+] | |
| 106.44 | |
| CHEBI:65242 | |
| ACS | |
| 2.5 g/cm3 at 20°C |
| 100g | |
| ClNaO3 | |
| sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal | |
| YZHUMGUJCQRKBT-UHFFFAOYSA-M | |
| sodium;chlorate | |
| 516902 | |
| 106.45 | |
| Odorless | |
| Sodium chlorate, For analysis ACS |
Safety and Handling
- Sodium chlorate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Oxidising solid Category 1
Hazard Statement
- H271-May cause fire or explosion; strong oxidiser.
- H302-Harmful if swallowed.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
- MIXTURE LIST-Contain: Sodium chlorate
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.