missing translation for 'onlineSavingsMsg'
Learn More

Sodium chlorate, ACS, 99.0% min, Alfa Aesar™

Maximum level of impurities: Insoluble matter 0.005%, BrO3 0.015%, Cl 0.005%, SO4 0.001%, Heavy metals (as Pb) 0.001%, Fe 5ppm, Ca 0.002%, Mg 0.002%, K 0.01%

Brand:  Alfa Aesar 044408.A3

Code : NEW

Additional Details : CAS Number : 7775-09-9 Weight : 2.00000kg

Quantity 2kg

 View more versions of this product

Product Code. 11468600

  • £201.30 / 2kg


Estimated Shipment date
from Supplier 19-06-2018
Add to basket

Specifications

Chemical Identifiers

Sodium chlorate
7775-09-9
Crystalline
NaClO3
MFCD00003479
For analysis ACS
ACS Grade
99% min.
106.45

Specifications

Sodium chlorate
7775-09-9
Crystalline
NaClO3
MFCD00003479
Colorless
For analysis ACS
ACS Grade
99% min.
106.45
Odorless
2kg
Videos
SDS