missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chlorate, ≥99%, Reagent Grade, Honeywell™

Product Code. 15672640 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15672640 1 kg Poly bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15672640 Supplier Honeywell Research Chemicals Supplier No. 2441471KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7775-09-9
Quantity 1 kg
Molecular Formula ClNaO3
Linear Formula NaClO3
MDL Number MFCD00003479
UN Number UN1495
Synonym sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
InChI Key YZHUMGUJCQRKBT-UHFFFAOYSA-M
SMILES [Na+].[O-][Cl](=O)=O
IUPAC Name sodium chlorate
Molecular Weight (g/mol) 106.44
PubChem CID 516902
ChEBI CHEBI:65242
Formula Weight 106.44g/mol
Percent Purity ≥99%
Grade ReagentPlus
Packaging Poly bottle
Chemical Name or Material Sodium Chlorate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.