missing translation for 'onlineSavingsMsg'
Learn More

Sodium chlorate, 99% min, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 014267.D9

Quantity 5 x 1kg

 View more versions of this product

Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11387857

  • ONLINE EXCLUSIVE PRICE
    £136.00
    £122.65 / 5kg
    Save £13.35
    9% Off
 Restricted Item
Estimated Shipment: 21-10-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

7775-09-9
106.44
YZHUMGUJCQRKBT-UHFFFAOYSA-M
516902
sodium;chlorate
ClNaO3
MFCD00003479
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
CHEBI:65242
[O-]Cl(=O)=O.[Na+]

Specifications

7775-09-9
99% min.
MFCD00003479
Soluble in water,glycerol,hydrazine,acetone and methanol. Slightly soluble in ethanol and ammonia.
[O-]Cl(=O)=O.[Na+]
106.44
CHEBI:65242
Odorless
2.5
5 x 1kg
ClNaO3
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
YZHUMGUJCQRKBT-UHFFFAOYSA-M
sodium;chlorate
516902
106.45
Colorless granules
Sodium chlorate
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.