missing translation for 'onlineSavingsMsg'
Learn More

Sodium chlorate, 99% min

Product Code. 11377857
Change view
Click to view available options
Quantity:
100 g
1 kg
5 x 1 kg
Unit Size:
100g
1kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377857 100 g 100g
11367857 1 kg 1kg
11387857 5 x 1 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11377857 Supplier Thermo Scientific Alfa Aesar Supplier No. 014267.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 7775-09-9 ∣ ClNaO3 ∣ 106.44 g/mol

Sodium chlorate is used to prepare chlorine dioxide in the pulp and paper industry. It is utilized for bleaching the wood pulp in order to make environmentally friendly and quality white paper products. It is also used to produce other perchlorate compounds in the industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-09-9
Molecular Formula ClNaO3
Molecular Weight (g/mol) 106.44
MDL Number MFCD00003479
InChI Key YZHUMGUJCQRKBT-UHFFFAOYSA-M
Synonym sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
PubChem CID 516902
ChEBI CHEBI:65242
IUPAC Name sodium chlorate
SMILES [Na+].[O-][Cl](=O)=O

Specifications

Melting Point 248°C
Color Colorless
Physical Form Granules
Quantity 100 g
Odor Odorless
Appearance Colorless granules
Assay Percent Range 99% min.
Density 2.5 g/cm3
Solubility Information Soluble in water,glycerol,hydrazine,acetone and methanol. Slightly soluble in ethanol and ammonia.
Formula Weight 106.45
Percent Purity ≥99%
Grade Ungraded
Chemical Name or Material Sodium chlorate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.