missing translation for 'onlineSavingsMsg'
Learn More

Sodium chlorate, 99% min, Alfa Aesar™

Brand:  Alfa Aesar 014267.A1

Code : NEW

Additional Details : CAS Number : 7775-09-9 Weight : 1.00000kg Transport : UN number : 1495 Chem class : 1 Pack group : II

Quantity 1kg

 View more versions of this product

Product Code. 11367857

  • £25.70 / 1kg


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Specifications

Chemical Identifiers

Sodium chlorate
Granular
NaClO3
MFCD00003479
7775-09-9
99% min.
106.45

Specifications

Sodium chlorate
Granular
NaClO3
106.45
2.5g/cm3
Colorless
7775-09-9
99% min.
Colorless granules
MFCD00003479
Odorless
1kg
Videos
SDS