missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloranilate trihydrate, 95%, Thermo Scientific™

Product Code. 10005790 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 10005790

Brand: Thermo Scientific Acros 419500250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 36275-66-8
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C6Cl2Na2O4·3H2O
MDL Number MFCD00150173
Quantity 25 g
Synonym chloranilic acid sodium salt, chloranilic acid disodium salt, disodium chloranilate, chloroanilic acid sodium salt, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro-3,6-dihydroxy-, disodium salt, sodium chloranilate, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro-3,6-dihydroxy-, sodium salt 1:2, disodium 2,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,4-diolate, chloranilic acid sodium salt hydrate, disodium 2,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,4-bis olate
InChI Key FITANBHHCLWRKG-UHFFFAOYSA-L
SMILES C1(=C(C(=O)C(=C(C1=O)Cl)[O-])Cl)[O-].[Na+].[Na+]
IUPAC Name disodium;2,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,4-diolate
Molecular Weight (g/mol) 307
PubChem CID 118938
Formula Weight 307
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Sodium chloranilate trihydrate, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.