missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Channel Pan Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577723
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Epithelial sodium channels are amiloride-sensitive members of the Degenerin/epithelial sodium channel (Deg/ENaC) superfamily of ion channels. Members of this superfamily of ion channels share organizational similarity in that they all possess two short intracellular amino and carboxyl termini, two short membrane spanning segments, and a large extracellular loop with a conserved cysteine-rich region. There are three homologous isoforms of the ENaC (alpha, beta, and gamma) protein. ENaC in the kidney, lung, and colon plays an essential role in trans-epithelial sodium and fluid balance. ENaC also mediates aldosterone-dependent sodium reabsorption in the distal nephron of the kidney, thus regulating blood pressure. ENaC is thought to be regulated, in part, through association with the cystic fibrosis transmembrane conductance regulator (CFTR) chloride ion channel. Gain-of-function mutations in beta- or gamma-ENaC can cause severe arterial hypertension (Liddels syndrome) and loss-of-function mutations in alpha- or beta-ENaC causes pseudohypoaldosteronism (PHA-1).
Specifications
| Sodium Channel Pan | |
| Polyclonal | |
| Unconjugated | |
| SCN1A | |
| SP-19 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 24766, 29701, 29710, 497770, 110880, 11280, 20273, 25722, 6323, 6328, 6329, 6336, 20271, 20274, 25665, 29571, 20265, 20269, 24046, 6326, 6331, 6334, 6335, 78956, 81574, 110876, 20264 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| A2APX8, Q9NY46, P04774, P35498, P35499, Q14524, Q9WTU3, Q9Y5Y9, O08562, Q62205, Q62968, Q15858, Q6QIY3, Q99250, Q9ER60, Q9R053, O88420, O88457, P04775, P15390, Q9JJV9, Q9UQD0, P08104, P15389, Q9UI33 | |
| Scn10a, Scn11a, Scn1a, Scn2a, Scn3a, Scn4a, Scn5a, Scn8a, Scn9a | |
| Peptide TEEQKKYYNAMKKLGSKK(C), corresponding to amino acid residues 1501-1518 of rat Nav 1.1 | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction