missing translation for 'onlineSavingsMsg'
Learn More

Sodium Channel inhibitor 1, MedChemExpress

Product Code. p-200101641
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499327 5 mg 5mg
16482857 50 mg 50mg
16436906 100 mg 100mg
16416788 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499327 Supplier Medchem Express Supplier No. HY15736

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sodium Channel inhibitor1, one of 3-Oxoisoindoline-1-carboxamides, is a novel and selective voltage-gated sodium channel for pain treatment.

Sodium Channel inhibitor1, one of 3-Oxoisoindoline-1-carboxamides, is a novel and selective voltage-gated sodium channel for pain treatment.

Specifications

Chemical Name or Material Sodium Channel inhibitor 1
CAS 1198117-23-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H19F4N3O3
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (211.23 mM; Need ultrasonic)
SMILES O=C(C1N(CCC2=NC=CC=C2)C(C3=C1C(F)=CC=C3)=O)NCC4=CC=C(OC(F)(F)F)C=C4
Molecular Weight (g/mol) 473.42
Formula Weight 473.42
Percent Purity 98.66%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.