missing translation for 'onlineSavingsMsg'
Learn More

Sodium carboxymethyl cellulose (Viscosity:5000-15000 mPa.s), MedChemExpress

Product Code. 16422728
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422728 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422728 Supplier Medchem Express Supplier No. HYY1889
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sodium carboxymethyl cellulose (Viscosity:5000-15000 mPa.s) is the sodium salt of cellulose arboxymethyl and frequently used as viscous agent, paste and barrier agent.

Sodium carboxymethyl cellulose (Viscosity:5000-15000 mPa.s) is the sodium salt of cellulose arboxymethyl and frequently used as viscous agent, paste and barrier agent.

Specifications

Chemical Name or Material Sodium carboxymethyl cellulose (Viscosity:5000-15000 mPa.s)
CAS 9004-32-4
Color Off-White
Purity Grade Notes Research
Quantity 5 g
Synonym CMC-Na (Viscosity:5000-15000 mPa.s)
SMILES [Sodium carboxymethyl cellulose]
Grade Research
Physical Form Solid

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.