missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate (White Crystalline Powder), Fisher BioReagents
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
Description
This sodium salt is used in buffers for molecular biology and cell culture applications.
Specifications
Specifications
| Boiling Point | 1600°C |
| Melting Point | 854°C |
| CAS | 497-19-8 |
| Color | White |
| pH | 11.3 |
| Physical Form | Solid |
| Quantity | 1 kg |
| Assay Percent Range | ≥99.5 % |
| Molecular Formula | CNa2O3 |
| MDL Number | MFCD00003494 |
| Show More |
Safety and Handling
Product Identifier
- Sodium carbonate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction