missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., anhydrous, ≥99.8% (calc. to the dried substance), Honeywell Fluka™

Product Code. 15673400 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
5 kg
6 x 500 g
25 kg
6 x 1 kg
Unit Size:
1kg
2.5kg
250g
25kg
500g
5kg
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15673400 25 kg 25kg
15663400 250 g 250g
15683400 500 g 500g
15643400 1 kg 1kg
15653400 2.5 kg 2.5kg
15613410 6 x 500 g Pack of 6
15693400 5 kg 5kg
15603410 6 x 1 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15673400 Supplier Honeywell-Fluka Supplier No. 3143225KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., anhydrous,≥99.8% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Physical Form Anhydrous
Quantity 25 kg
Linear Formula Na2CO3
UN Number NONH for all modes of transport
Beilstein 4154566
IUPAC Name disodium;carbonate
Formula Weight 105.99g/mol
Percent Purity ≥99.8% (calc. to the dried substance)
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium Carbonate, Reag. Ph. Eur. reag. ISO
compliance-icons
Product Identifier
  • Sodium carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.