missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate, Puriss., meets analytical spec. of BP, Ph. Eur., NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance), Honeywell Fluka™
BIOPHARMA

Product Code. 15631300 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
25 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
25kg
5kg
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15631300 25 kg 25kg
15611300 1 kg 1kg
15621300 2.5 kg 2.5kg
15641300 5 kg 5kg
15651300 6 x 1 kg Pack of 6
15661300 6 x 2.5 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15631300 Supplier Honeywell-Fluka Supplier No. 1341825KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Melting Point 851°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Quantity 25 kg
Assay Percent Range 99.5 to 100.5%
Linear Formula Na2CO3
UN Number NONH for all modes of transport
Beilstein 4154566
IUPAC Name disodium carbonate
Formula Weight 105.99
Percent Purity 99.5 to 100.5% (calc. to the dried substance)
Grade puriss.
Packaging Paper / Poly Bag
Density 2.53 g/cm3
Chemical Name or Material Sodium carbonate, Puriss., meets analytical specification of BP, Ph. Eur., NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.