missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate, Powder, ≥99.5%, ACS reagent, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
12 kg
Packaging:
Fiberboard Box
Poly Bottle
Unit Size:
12kg
1kg
2.5kg
500g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Melting Point | 851°C (literature) |
| CAS Min % | ≥90.0000% |
| CAS Max % | ≤100.0000% |
| Physical Form | Powder |
| Quantity | 2.5 kg |
| Linear Formula | Na2CO3 |
| Beilstein | 4154566 |
| IUPAC Name | disodium carbonate |
| Formula Weight | 105.99 |
| Percent Purity | >99.5% |
| Show More |
Safety and Handling
Product Identifier
- Sodium carbonate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : sodium carbonate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction