missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, Monohydrate, Crystal, N.F. - F.C.C., J.T. Baker™

Product Code. 15568144 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15568144 500 g Poly Bottle 500g
15578144 2.5 kg Poly Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15568144 Supplier Avantor J.T.Baker Supplier No. 360001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5968-11-6
Molecular Formula CH2Na2O4
Molecular Weight (g/mol) 124.00
InChI Key MQRJBSHKWOFOGF-UHFFFAOYSA-L
Synonym sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop
PubChem CID 2735133
IUPAC Name disodium;carbonate;hydrate
SMILES C(=O)([O-])[O-].O.[Na+].[Na+]

Specifications

Physical Form Crystals
Quantity 500 g
Formula Weight 124
Grade FCC/NF
Packaging Poly Bottle
Specific Gravity 1L = 2.25kg
Chemical Name or Material Sodium Carbonate, Monohydrate
compliance-icons
Product Identifier
  • Sodium carbonate monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sodium carbonate monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.