Learn More
Sodium carbonate monohydrate, ACS, 99.5% min
Description
It is widely employed as a water softener, an electrolyte, as a good conductor in electrolysis, as a primary standard for acid-base titrations and mostly as a buffer component in applications such as chromatography, capillary electrophoresis and enzyme catalysis. Also used in glass manufacture, detergent manufacture, sodium chemicals, carbonate chemicals manufacture, pulp and paper, brine treatment, flue gas desulfurization, coal treatment and ion exchange resin regeneration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 100°C (in 2H2O) |
| Color | Colorless to White |
| Physical Form | Granules |
| Quantity | 2 kg |
| Merck Index | 14,8596 |
| Solubility Information | Soluble in water and glycerol. Insoluble in alcohol,acetone,alkyl acetates,benzonitrile and liquid ammonia. |
| Formula Weight | 124.01 (105.99 Anhydrous) |
| Percent Purity | ≥99.5% |
| Grade | ACS |
| Odor | Odorless |
| Show More |
Safety and Handling
- Sodium carbonate monohydrate, ACS
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: Carbonic acid, disodium salt, monohydrate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.