missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate monohydrate, 99.6%, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-3735137
29.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
5968-11-6
CNa2O3·H2O
124.01
MFCD00149177
MQRJBSHKWOFOGF-UHFFFAOYSA-L
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop
2735133
disodium;carbonate;hydrate
C(=O)([O-])[O-].O.[Na+].[Na+]
5968-11-6
CNa2O3·H2O
124.01
MFCD00149177
MQRJBSHKWOFOGF-UHFFFAOYSA-L
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop
2735133
disodium;carbonate;hydrate
C(=O)([O-])[O-].O.[Na+].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 851.0°C
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 99.6%
Linear Formula Na2CO3·H2O
Fieser 07,322
Merck Index 15, 8731
Solubility Information Solubility in water: 210g/L. Other solubilities: soluble in 1.8 parts boiling water, in 7 parts glyceral, insoluble in alcohol
Formula Weight 124.01
Percent Purity 99.6%
Grade ACS Reagent
Loss on Drying 13.0% to 15.0% (150°C)
Packaging Plastic bottle
Chemical Name or Material Sodium carbonate monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium carbonate monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'