missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate Monohydrate, ≥99.5%, ACS Reagent, Honeywell™ Fluka™

Product Code. 15642300 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15642300 100 g Poly Bottle 100g
15662300 500 g Poly Bottle 500g
15652300 2.5 kg Poly Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15642300 Supplier Honeywell-Fluka Supplier No. 230952100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5968-11-6
Molecular Formula CH2Na2O4
Molecular Weight (g/mol) 124.00
MDL Number MFCD00149177
InChI Key MQRJBSHKWOFOGF-UHFFFAOYSA-L
Synonym sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop
PubChem CID 2735133
IUPAC Name disodium;carbonate;hydrate
SMILES C(=O)([O-])[O-].O.[Na+].[Na+]

Specifications

Melting Point 851°C
Color White
Quantity 100 g
Linear Formula Na2CO3 · H2O
UN Number NONH for all modes of transport
Formula Weight 124g/mol
Percent Purity ≥99.5%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder
Density 2.25g/cm3
Chemical Name or Material Sodium Carbonate Monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.