missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate monohydrate, 85% min

Product Code. 11347707
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347707 5 kg 5kg
11357707 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347707 Supplier Thermo Scientific Alfa Aesar Supplier No. 014108.A7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5968-11-6 | CH2Na2O4 | 124.00 g/mol

Sodium carbonate monohydrate is widely employed as a water softener, an electrolyte, as a good conductor in electrolysis, as a primary standard for acid-base titrations and mostly as a buffer component in applications such as chromatography, capillary electrophoresis and enzyme catalysis. Also used in glass manufacture, detergent manufacture, sodium chemicals, carbonate chemicals manufacture, pulp and paper, brine treatment, flue gas desulfurization, coal treatment and ion exchange resin regeneration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5968-11-6
Molecular Formula CH2Na2O4
Molecular Weight (g/mol) 124.00
MDL Number MFCD00149177
InChI Key MQRJBSHKWOFOGF-UHFFFAOYSA-L
Synonym sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop
PubChem CID 2735133
IUPAC Name disodium;carbonate;hydrate
SMILES C(=O)([O-])[O-].O.[Na+].[Na+]

Specifications

Melting Point 100°C (in -H2O)
Color Colorless to White
Physical Form Crystalline
Quantity 5 kg
Merck Index 14,8596
Solubility Information Soluble in water; Insoluble in alcoholSoluble in water and glycerol; Insoluble in alcohol,acetone,alkyl acetates,benzonitrile and liquid ammonia.
Formula Weight 124.01 (105.99 Anhydrous)
Percent Purity ≤85%
Odor Odorless
Sensitivity Hygroscopic
Density 2.250 g/mL
Chemical Name or Material Sodium carbonate monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium carbonate monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sodium carbonate monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.