missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate Decahydrate, Technical, Fisher Chemical

Product Code. 11473703 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
3 kg
Packaging:
Plastic powder jar
Unit Size:
3kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11473703 3 kg Plastic powder jar 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11473703 Supplier Fisher Chemical Supplier No. S/2700/63
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

Melting Point 34°C
CAS 2-1-6132
Color White
pH 11 to 12
Physical Form Solid
Quantity 3 kg
Molecular Formula CH20Na2O13
MDL Number 149178
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]
IUPAC Name disodium;carbonate;decahydrate
Molecular Weight (g/mol) 286.14
PubChem CID 151402
Formula Weight 286.01g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Sodium carbonate decahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.