Learn More
Sodium carbonate decahydrate, 99+%
Description
Sodium carbonate decahydrate is used as a raw material in various industries like glass, paper making, soap, detergent, textile and leather. It is also used as a food additive. It is also involved as a precipitation agent of carbonate as well as an analytical reagent in laboratories. It plays an important role as strong base and powerful electrolyte. It finds application in metal refining and cement production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium carbonate decahydrate is used as a raw material in various industries like glass, paper making, soap, detergent, textile and leather. It is also used as a food additive. It is also involved as a precipitation agent of carbonate as well as an analytical reagent in laboratories. It plays an important role as strong base and powerful electrolyte. It finds application in metal refining and cement production.
Solubility
Soluble in water and glycerol. Insoluble in alcohol.
Notes
Air sensitive. Incompatible with strong acids and aluminum. Keep the container tightly closed.
Specifications
Specifications
| Melting Point | 32°C to 34°C |
| Quantity | 10,000 g |
| Linear Formula | Na2CO3·10H2O |
| Merck Index | 14,8596 |
| Solubility Information | Soluble in water and glycerol. Insoluble in alcohol. |
| Formula Weight | 286.14 (105.99 Anhydrous) |
| Percent Purity | ≥99% |
| Density | 1.46 |
| Chemical Name or Material | Sodium carbonate decahydrate |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.