missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate decahydrate, 99+%, for analysis, ACROS Organics™

Brand:  Acros Organics 213500010

Code : 29

Additional Details : CAS Number : 6132-02-1 Weight : 1.05000kg

Quantity 1kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 10348110

  • £31.10 / 1kg


EU Stock 19
Add to basket

Specifications

Chemical Identifiers

Sodium carbonate decahydrate
6132-02-1
≥99.0%
CNa2O3·10H2O
286.14
p.a.
For analysis
99+%
Na2CO3·10H2O
MFCD00149178

Specifications

Sodium carbonate decahydrate
6132-02-1
For analysis
≥99.0%
CNa2O3·10H2O
286.14
07,322
20ppm max.
1.46g/mL
2ppm max.
Plastic bottle
0.002% max.
White
p.a.
5ppm max.
Small Crystals
99+%
Na2CO3·10H2O
MFCD00149178
15, 8731
5ppm max.
10ppm max.
2ppm max.
100ppm max.
0.0005% max.
1kg
Videos
SDS