missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate, Anhydrous, Granular, N.F. - F.C.C., J.T. Baker™
532.00 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle
Specifications
Physical Form | Granules |
Quantity | 3 kg |
Formula Weight | 105.99 |
Grade | FCC/NF |
Packaging | Poly Pail |
Specific Gravity | 1L = 2.53kg |
Chemical Name or Material | Sodium Carbonate, Anhydrous |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'