missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate Anhydrous, Extra Pure, SLR, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
2.5kg
500g
5kg
Description
Sodium Carbonate Anhydrous, Extra Pure, SLR
Specifications
Specifications
| Boiling Point | 1600°C |
| Melting Point | 854°C |
| Color | White |
| pH | 11.3 |
| Physical Form | Solid |
| Quantity | 2.5 kg |
| IUPAC Name | disodium;carbonate |
| Formula Weight | 105.99g/mol |
| Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Sodium carbonate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction