missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate Anhydrous, Certified AR for Analysis, Fisher Chemical™
146.26 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
Sodium Carbonate Anhydrous, Certified AR for Analysis
Specifications
Boiling Point | 1600°C |
Melting Point | 854°C |
Color | White |
pH | 11.3 |
Physical Form | Solid |
Quantity | 2.5 kg |
IUPAC Name | disodium carbonate |
Formula Weight | 105.99g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Sodium carbonate anhydrous
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'