missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate, ACS Reagent, anhydrous, powder or granules, ≥99.5%, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
12 kg
4 x 2.5 kg
Packaging:
Drum With Inliner
Fiberboard Box
Poly Bottle
Unit Size:
12kg
1kg
2.5kg
500g
5kg
Pack of 4
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Melting Point | 851°C (literature) |
| CAS Min % | ≥90.0000% |
| CAS Max % | ≤100.0000% |
| Physical Form | Powder/Granules |
| Quantity | 500 g |
| Linear Formula | Na2CO3 |
| Beilstein | 4154566 |
| IUPAC Name | disodium carbonate |
| Formula Weight | 105.99 |
| Percent Purity | >99.5% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction