missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate, 99+%, for analysis, granular
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
Packaging:
Plastic Bottle
Plastic Drum
Unit Size:
1kg
2.5kg
5kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifications
Specifications
| Boiling Point | 1600.0°C |
| Melting Point | 851.0°C |
| Color | White |
| Physical Form | Fine Granular Powder |
| Quantity | 5 kg |
| Assay Percent Range | 99+% |
| Linear Formula | Na2CO3 |
| Fieser | 7332 |
| Merck Index | 15, 8731 |
| Solubility Information | Solubility in water: 22 g/100mL (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol |
| Show More |
Safety and Handling
Product Identifier
- Sodium carbonate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction