missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium-Calcium Exchanger Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69515
This item is not returnable.
View return policy
Description
Sodium-Calcium Exchanger Polyclonal specifically detects Sodium-Calcium Exchanger in Human samples. It is validated for Western Blot.
Specifications
| Sodium-Calcium Exchanger | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Na(+)/Ca(2+)-exchange protein 3, Na+/Ca2+ exchanger 3, NCX3sodium/calcium exchanger SLC8A3, sodium/calcium exchanger 3, solute carrier family 8 (sodium/calcium exchanger), member 3, solute carrier family 8 (sodium-calcium exchanger), member 3 | |
| Rabbit | |
| 103 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q96QG2 | |
| SLC8A3 | |
| Synthetic peptides corresponding to SLC8A3(solute carrier family 8 (sodium-calcium exchanger), member 3) The peptide sequence was selected from the N terminal of Sodium-Calcium Exchanger. Peptide sequence SAPEILLSLIEVCGHGFIAGDLGPSTIVGSAAFNMFIIIGICVYVIPDGE. | |
| Affinity purified | |
| RUO | |
| 6547 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction