missing translation for 'onlineSavingsMsg'
Learn More

Sodium Butyrate, TRC

Product Code. 30137668
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137668 50 g 50g
30137321 100 g 100g
30006865 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137668 Supplier Toronto Research Chemicals Supplier No. TRCS61517550G
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-54-7
Molecular Formula C4 H7 O2 . Na
Molecular Weight (g/mol) 110.09
Synonym Butanoic acid, sodium salt (1:1), Butanoic acid, sodium salt (9CI), Butyric acid, sodium salt (6CI,8CI), Adimix, Butyrate sodium, Gustor XXI B 70, Sodium butanoate, Sodium butyrate, Sodium n-butyrate, Sodium propanecarboxylate
IUPAC Name sodium;butanoate
SMILES [Na+].CCCC(=O)[O-]

Specifications

Quantity 50 g
InChI Formula InChI=1S/C4H8O2.Na/c1-2-3-4(5)6;/h2-3H2,1H3,(H,5,6);/q;+1/p-1
Formula Weight 110.0344
Chemical Name or Material Sodium Butyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.