missing translation for 'onlineSavingsMsg'
Learn More

Sodium butyrate, 98+%

Product Code. 11448943
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448943 500 g 500g
11423980 5 g 5g
11438943 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448943 Supplier Thermo Scientific Alfa Aesar Supplier No. A11079.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 156-54-7 | C4H7NaO2 | 110.09 g/mol

Decreases calcium release from intracellular stores. Inhibits histone deacetylase.Sodium butyrate is used in the treatment of human spinal muscular atrophy patients. Further, it is used in the determination of the role of histone acetylation in chromatin structure and function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decreases calcium release from intracellular stores. Inhibits histone deacetylase.Sodium butyrate is used in the treatment of human spinal muscular atrophy patients. Further, it is used in the determination of the role of histone acetylation in chromatin structure and function.

Solubility
Soluble in water, dimethyl sulfoxide and ethanol. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-54-7
Molecular Formula C4H7NaO2
Molecular Weight (g/mol) 110.09
MDL Number MFCD00002816
InChI Key MFBOGIVSZKQAPD-UHFFFAOYSA-M
Synonym sodium butyrate, sodium butanoate, butyric acid sodium salt, butanoic acid, sodium salt, butyrate sodium, butyric acid, sodium salt, sodium n-butyrate, sodium propanecarboxylate, butyric acid, na, butanoic acid, sodium salt 1:1
PubChem CID 5222465
ChEBI CHEBI:64103
SMILES [Na+].CCCC([O-])=O

Specifications

Melting Point >300°C
Assay Percent Range >98%
Quantity 500 g
Beilstein 3629439
Sensitivity Hygroscopic
Solubility Information Soluble in water,dimethyl sulfoxide and ethanol. Insoluble in ether.
IUPAC Name sodium butanoate
Formula Weight 110.09
Chemical Name or Material Sodium butyrate
What is the solubility of Sodium butyrate, 98+%?

Sodium butyrate is soluble in water (0.1 g/mL), DMSO (<1 mg/mL), ethanol (~5 mg/mL), and PBS pH7. 2 (~10 mg/mL). Sodium butyrate is insoluble in ether.

compliance-icons
Product Identifier
  • Sodium butyrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Butanoic acid, sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.